Anti-CD52 Recombinant Antibody [Clone: YTH 34.5-G2b (Campath-1G)]
Catalog # PRSI24-193
Supplier: ProSci Inc.
New Product
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:CD52 molecule
- Antigen-Symbol:CD52
- Klonalität:Recombinant
- Klon:YTH 34.5-G2b (Campath-1G)
- Konjugation:Unconjugated
- ELISA:Yes
- Durchflusszytometrie:Yes
- ImmunoChemie:Yes
- Isotyp:IgG1 kappa
- Reaktivität:Cynomolgus monkey,Rhesus monkey,Human
- Western blot:Yes
- Kreuzadsorption:No
- Form:Liquid
- Gen-ID:1043
- Antigen-Synonyme:Cambridge pathology 1 antigen|CDw52|Epididymal secretory protein E5|Human epididymis-specific protein 5
- Modification:Unmodified
- Lagerungspuffer:PBS with 0,02% Proclin 300
- Lagertemperatur:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Transporttemperatur:Shipped on blue ice
- Immunogen:Human peripheral mononuclear cells enriched for T-cells.
- Validierte Anwendungen:Cytotoxicity Assay,in vivo
- Größe:0,2 mg
- Packung:Vial
- VE:0,2 mg
Specifications
About this item
Type: Primary
Antigen: CD52 molecule
Clonality: Recombinant
Clone: YTH 34.5-G2b (Campath-1G)
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Human;Rhesus Monkey;Cynomolgus Monkey