Anti-V5 Epitope Tag Recombinant Antibody [Clone: SV5-P-K]
PRSI24-171
New Product
- Antikörper-Typ:Primär
- Klonalität:Recombinant
- Klon:SV5-P-K
- Konjugation:Unconjugated
- ELISA:Yes
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Immunpräzipitation:Yes
- Isotyp:IgG2b kappa
- Reaktivität:Paramyxovirus simian virus 5
- Western blot:Yes
- Kreuzadsorption:No
- Form:Liquid
- Antigen-Synonyme:V5 epitope tag|GKPIPNPLLGLDST epitope tag|PK tag|protein rev|Regulator of expression of viral proteins|PK-tag|GKPIPNPLLGLDST tag
- Modification:Unmodified
- Lagerungspuffer:PBS with 0,02% Proclin 300
- Lagertemperatur:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Transporttemperatur:Shipped on blue ice
- Immunogen:Pk (V5) Epitope Tag (GKPIPNPLLGLDST).
- Validierte Anwendungen:RIA
- Größe:0,2 mg
- Packung:Vial
- VE:0,2 mg
Type: Primary
Antigen: V5 Epitope Tag
Clonality: Recombinant
Clone: SV5-P-K
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG2b kappa
Reactivity: Paramyxovirus Simian Virus 5