Anti-FNBP4 Rabbit Polyclonal Antibody
PRSI15-210
New Product
- Antikörper-Typ:Primär
- Antigen-Name:Formin binding protein 4
- Antigen-Symbol:FNBP4
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:23360
- Antigen-Synonyme:FBP3formin-binding protein 4|formin-binding protein 30
- Modification:None
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Lagertemperatur:Store at −20 °C. Avoid freeze/thaw cycles
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human FNBP4 (NP_056123.2)
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Type: Primary
Antigen: FNBP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse