Anti-Rad51D Rabbit Polyclonal Antibody
PRSI23-011
New Product
- Antikörper-Typ:Primär
- Antigen-Name:RAD51 paralog D
- Antigen-Symbol:Rad51D
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:5892
- Antigen-Synonyme:HsTRAD|RAD51L3|DNA repair protein RAD51 homolog 4|RAD51-like 3 (S. cerevisiae)|TRAD|recombination repair protein|R51H3|RAD51D
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Lagertemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-328 of human Rad51D (NP_002869.3)
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Type: Primary
Antigen: Rad51D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat