Anti-RAC3 Rabbit Polyclonal Antibody
PRSI22-986
New Product
- Antikörper-Typ:Primär
- Antigen-Name:Rac family small GTPase 3
- Antigen-Symbol:RAC3
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:5881
- Antigen-Synonyme:small GTP binding protein Rac3|p21-Rac3|small GTP binding protein Rac3)|rho family|ras-related C3 botulinum toxin substrate 3|ras-related C3 botulinum toxin substrate 3 (rho family
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Lagertemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-192 of human RAC3 (NP_005043.1)
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Type: Primary
Antigen: RAC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat