Anti-HNRNPA1 Rabbit Polyclonal Antibody
Artikelnr. PRSI22-980
Lieferant: ProSci Inc.
New Product
Spezifikationen
- Antikörper-Typ:Primär
- Antigen-Name:Heterogeneous nuclear ribonucleoprotein A1
- Antigen-Symbol:HNRNPA1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Western blot:Yes
- Form:Liquid
- Gen-ID:3178
- Antigen-Synonyme:ALS20|HNRPA1|hnRNP-A1|hnRNP A1|ALS19|IBMPFD3|HNRPA1L3
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Lagertemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-372 of human HNRNPA1 (NP_112420.1)
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Spezifikationen
Über diesen Artikel
Type: Primary
Antigen: HNRNPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat