Anti-CNDP1 Rabbit Polyclonal Antibody
Catalog # PRSI22-976
Supplier: ProSci Inc.
New Product
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Beta-Ala-His dipeptidase
- Antigen-Symbol:CNDP1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:84735
- Antigen-Synonyme:CN1|CNDP dipeptidase 1|Carnosine dipeptidase 1|CPGL2|CNDP1|Beta-Ala-His dipeptidase|Serum carnosinase|Glutamate carboxypeptidase-like protein 2
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Lagertemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human CNDP1 (NP_116038.4)
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Specifications
About this item
Type: Primary
Antigen: CNDP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat