Anti-OSGEP Rabbit Polyclonal Antibody
Artikelnr. PRSI22-965
Lieferant: ProSci Inc.
New Product
Spezifikationen
- Antikörper-Typ:Primär
- Antigen-Name:O-sialoglycoprotein endopeptidase
- Antigen-Symbol:OSGEP
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:55644
- Antigen-Synonyme:GAMOS3|tRNA threonylcarbamoyladenosine biosynthesis protein OSGEP|probable O-sialoglycoprotein endopeptidase|t(6)A synthase|OSGEP|probable tRNA threonylcarbamoyladenosine biosynthesis protein OSGEP|t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEP|TCS3|hOSGEP|KAE11|probable tRNA N6-adenosine threonylcarbamoyltransferase|GCPL1|N6-L-threonylcarbamoyladenine synthase|PRSMG1
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Lagertemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-335 of human OSGEP (NP_060277.1)
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Spezifikationen
Über diesen Artikel
Type: Primary
Antigen: OSGEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat