Anti-ERGIC2 Rabbit Polyclonal Antibody
PRSI22-894
New Product
- Antikörper-Typ:Primär
- Antigen-Name:ERGIC and golgi 2
- Antigen-Symbol:ERGIC2
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- Isotyp:IgG
- Western blot:Yes
- Form:Liquid
- Gen-ID:51290
- Antigen-Synonyme:MGC111152|ERGIC2|cd002|Erv41|CDA14|PTX1
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Lagertemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 55-310 of human ERGIC2 (NP_057654.2)
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Type: Primary
Antigen: ERGIC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse