Anti-MAP1LC3B Rabbit Polyclonal Antibody
PRSI22-775
New Product
- Antikörper-Typ:Primär
- Antigen-Name:Microtubule-associated protein 1 light chain 3 beta
- Antigen-Symbol:MAP1LC3B
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Reaktivität:Pig,Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:81631
- Antigen-Synonyme:MAP1A/1BLC3|Autophagy-related protein LC3 B|MAP1LC3B Antibody: LC3B|MAP1LC3B-a|MAP1ALC3|ATG8F
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Lagertemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:A synthetic peptide of human LC3B
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Type: Primary
Antigen: MAP1LC3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig