Anti-XRCC5 Rabbit Polyclonal Antibody
Catalog # PRSI19-904
Supplier: ProSci Inc.
New Product
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining)
- Antigen-Symbol:XRCC5
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Immunpräzipitation:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:7520
- Antigen-Synonyme:NFIV|KUB2|Ku86|KARP1|KARP-1|XRCC5|KU80
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Lagertemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 543-732 of human XRCC5 (NP_066964.1).
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Specifications
About this item
Type: Primary
Antigen: XRCC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat