Anti-MRPS30 Rabbit Polyclonal Antibody
PRSI19-893
New Product
- Antikörper-Typ:Primär
- Antigen-Name:Mitochondrial ribosomal protein S30
- Antigen-Symbol:MRPS30
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Immunpräzipitation:Yes
- Isotyp:IgG
- Western blot:Yes
- Form:Liquid
- Gen-ID:10884
- Antigen-Synonyme:PDCD9|MRP-S3PAP|28S ribosomal protein S3mitochondrial|mitochondrial large ribosomal subunit protein mS3programmed cell death 9|mitochondrial large ribosomal subunit protein mL65|S30mt|39S ribosomal protein S3mitochondrial
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Lagertemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 190-439 of human MRPS30 (NP_057724.2).
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Type: Primary
Antigen: MRPS30
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat