Anti-GOT1 Rabbit Polyclonal Antibody
PRSI19-881
New Product
- Antikörper-Typ:Primär
- Antigen-Name:Glutamic-oxaloacetic transaminase 1, soluble
- Antigen-Symbol:GOT1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Western blot:Yes
- Form:Liquid
- Gen-ID:2805
- Antigen-Synonyme:cAspAT|cCAT|ASTQTL1|GOT1|GIG18
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Lagertemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 244-413 of human GOT1 (NP_002070.1).
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Type: Primary
Antigen: GOT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat