Anti-BTAF1 Rabbit Polyclonal Antibody
Catalog # PRSI19-872
Supplier: ProSci Inc.
New Product
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:B-TFIID TATA-box binding protein associated factor 1
- Antigen-Symbol:BTAF1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- Immunpräzipitation:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- ChIP:Yes
- Form:Liquid
- Gen-ID:9044
- Antigen-Synonyme:BTAF1 RNA polymerase II|S. cerevisiae)|B-TFIID transcription factor-associated|B-TFIID transcription factor-associated 170 kDa subunit|TAFII17TATA-binding protein-associated factor 172|MOT1|TAF(II)17TAF172|TBP-associated factor 172|170kDa (Mot1 homolog|Mot1 homolog|ATP-dependent helicase BTAF1
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Lagertemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1600-1849 of human BTAF1 (NP_003963.1).
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Specifications
About this item
Type: Primary
Antigen: BTAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat