ContactUsLinkComponent
Human Recombinant BID (from E. coli)
  PRSI91-089
New Product
 :  
Human Recombinant BID (from E. coli)
  PRSI91-089
New Product
 :  
  • VE:
    0,05 mg
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Human
  • Lagerbedingungen:
    Store at −20 °C, stable for 6 months after receipt. Please minimise freeze-thaw cycles.
  • Endotoxingehalt:
    <0,1 ng/µg (1 IEU/µg) as determined by LAL test
  • Proteinsynonym:
    BH3-Interacting Domain Death Agonist|BID|p22 BID
  • Protein/Peptid-Name:
    BID
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    21,99 kDa
  • Sequence:
    Met1-Asp195
  • Zusammensetzung:
    Supplied as a 0,2 µm filtered solution of 20 mM PB, 100 mM KCl, pH 7,4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 µg/ml. Dissolve the Lyophilised protein in ddH₂O. Please aliquot the reconstituted solution to minimise freeze-thaw cycles.
  • Validierte Anwendungen:
    For biological assays
BH3-Interacting Domain Death Agonist (BID) is a member of the Bcl-2 protein family which regulates outer mitochondrial membrane permeability. BID is a pro-apoptotic member that causes cytochrome c to be released from the mitochondria intermembrane space into the cytosol. Interaction of Bid with Bak causes altered mitochondrial membrane permeability. BID contains only the BH3 domain, which is required for its interaction with the Bcl-2 family proteins and for its pro-death activity. BID is susceptible to proteolytic cleavage by caspases, calpains, Granzyme B and cathepsins. It is an integrating key regulator of the intrinsic death pathway that amplifies caspase-dependent and caspase-independent execution of neuronal apoptosis. Therefore pharmacological inhibition of BID provides a promising therapeutic strategy in neurological diseases where programmed cell death is prominent, and also offer a new strategy for the treatment of acute renal failure associated with ischemia-reperfusion. BID receives direct inputs from a key regulator of the cell cycle arrest/DNA repair machinery (ATM), and therefore is an excellent candidate to coordinate genotoxic stress responses and apoptotic cell death. BID is a novel pro-apoptosis Bcl-2 family protein that is activated by caspase 8 in response to Fas/TNF-R1 death receptor signals. Deletion of BID inhibits carcinogenesis in the liver, although this genetic alteration promotes tumorigenesis in the myeloid cells. This is likely related to the function of BID to promote cell cycle progression into S phase. BID could be also involved in the maintenance of genomic stability by engaging at mitosis checkpoint.
This recombinant protein can be used for biological assays.
 : For research use only.