Anti-DPT Rabbit Polyclonal Antibody
Catalog # PRSI56-560
Supplier: ProSci Inc.
New Product
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Dermatopontin
- Antigen-Symbol:DPT
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- Western blot:Yes
- Epitop:Amino acid range = 45-72
- Form:Liquid
- Gen-ID:Q07507
- Modification:None
- Lagerungspuffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:24 kDa
- Lagertemperatur:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Konzentration:2 mg/ml
- Transporttemperatur:4 °C
- Immunogen:This DPT antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 45-72 amino acids from the N-terminal region of human DPT
- Reinigung:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- VE:400 µl
Specifications
About this item
Dermatopontin is an extracellular matrix protein with possible functions in cell-matrix interactions and matrix assembly. The protein is found in various tissues and many of its tyrosine residues are sulphated. Dermatopontin is postulated to modify the behavior of TGF-beta through interaction with decorin. [provided by RefSeq].
- For WB starting dilution is: 1:1000
- For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: DPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human