Order Entry
Austria
ContactUsLinkComponent
Anti-IL4 Rat Monoclonal Antibody [clone: MP4-25D2]
Anti-IL4 Rat Monoclonal Antibody [clone: MP4-25D2]
  PRSI77-066
New Product
Anti-IL4 Rat Monoclonal Antibody [clone: MP4-25D2]
  PRSI77-066
New Product

 

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    interleukin 4
  • Antigen-Symbol:
    IL4
  • Klonalität:
    Monoklonal
  • Klon:
    MP4-25D2
  • Konjugation:
    Unconjugated
  • Wirt:
    Rat
  • Isotyp:
    IgG1 kappa
  • Kreuzadsorption:
    No
  • Neutralisation:
    Yes
  • Form:
    liquid
  • Gen-ID:
    P05112
  • Antigen-Synonyme:
    IL4
  • Lagerungspuffer:
    Phosphate-buffered aqueous solution, ph 7,2.
  • Lagertemperatur:
    The product should be stored undiluted at 4 °C. Do not freeze.
  • Transporttemperatur:
    4 °C
  • Validierte Anwendungen:
    Functional
  • Reinigung:
    The monoclonal antibody was purified utilizing affinity chromatography.
  • VE:
    0,5 mg

 

 

The MP4-25D2 monoclonal antibody specifically reacts with human Interleukin-4 (IL-4), expressed by the activated T cells and mast cells. IL-4 is species-specific and stimulates the proliferation and differentiation of B lymphocytes. It upregulates the expression of IgE receptors and class II MHC antigen. The MP4-25D2 is a neutralizing antibody and is reported to be cross-reactive with rhesus monkeys.

Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone: MP4-25D2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Human