Anti-EIF4E Rabbit Polyclonal Antibody
Vial
Artikelnr. PRSI18-547
Anbieter: ProSci Inc.
Springe zu:
Product Details & Documents
Type: Primary
Antigen: EIF4E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Spezifikationen
- Packung:Vial
- Western blot:Yes
- Modification:None
- Antikörper-Typ:Primär
- Konjugation:Unconjugated
- Form:Liquid
- Reinigung:Affinity purification
- Transporttemperatur:20
- Reaktivität:Human|Rat|Mouse
- Konzentration:Batch dependent
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Cat. No.:PRSI18-547
- Antigen-Name:Eukaryotic translation initiation factor 4E
- Lagertemperatur:Store at −20 °C. Avoid freeze/thaw cycles.
- Gen-ID:1977
- Immunpräzipitation:Yes
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 1-100 of human EIF4E (NP_001959.1)
- Antigen-Synonyme:Eukaryotic translation initiation factor 4E|eIF-4F 25 kDa subunit|EIF4EL1|eIF-4E|EIF4E|eIF4E|EIF4F|mRNA cap-binding protein
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Antigen-Symbol:EIF4E
- Klonalität:Polyklonal
- Isotyp:IgG
- Wirt:Rabbit



