Anti-MAN2B1 Rabbit Polyclonal Antibody
Catalog # PRSI23-985
Supplier: ProSci Inc.
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:mannosidase alpha class 2B member 1
- Antigen-Symbol:MAN2B1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:4125
- Antigen-Synonyme:lysosomal|mannosidase alpha-B|lysosomal alpha-mannosidase|LAMAN|lysosomal acid alpha-mannosidase|mannosidase|alpha B|MANB
- Modification:None
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Lagertemperatur:Store at −20 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 640-850 of human MAN2B1 (NP_000519.2)
- Reinigung:Affinity purification.
- Packung:Vial
- VE:50 µl
Specifications
About this item
Type: Primary
Antigen: MAN2B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse