Anti-GUCA2A Rabbit Polyclonal Antibody
Artikelnr. PRSI23-921
Lieferant: ProSci Inc.
Spezifikationen
- Antikörper-Typ:Primär
- Antigen-Name:Guanylin
- Antigen-Symbol:GUCA2A
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:2980
- Antigen-Synonyme:Guanylin|GUCA2|Guanylate cyclase-activating protein 1|Guanylate cyclase-activating protein I|Guanylate cyclase activator 2A|GCAP-I|HMW-guanylin|GUCA2A
- Modification:None
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Lagertemperatur:Store at −20 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 22-115 of human GUCA2A (NP_291031.2)
- Reinigung:Affinity purification.
- Packung:Vial
- VE:50 µl
Spezifikationen
Über diesen Artikel
Type: Primary
Antigen: GUCA2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat