3-Fluorstyrol
Glass bottle for solids, 5 g
Artikelnr. MOLE24155756-5G
Marketsource
Anbieter: Molekula
Springe zu:
Product Details & Documents
- CAS Nummer:350-51-6
- Formel:C₈H₇F
- MDL-Nummer:MFCD00000341
- Molekulargewicht:122.14 g/mol
- Siedepunkt:30 °C (1013 hPa)
- Dichte:1,025 g/cm³ (20 °C)
- :Density: 1.025 g/cm³ (20 °C)
- EINECS:206-504-9
Spezifikationen
Spezifikationen
- Packung:Glass bottle for solids
- VE:5 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



