Order Entry
Austria
ContactUsLinkComponent
Oxatomide ≥99% (durch HPLC)
Oxatomide ≥99% (durch HPLC)
Catalog # ENZOBMLEI1140010
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Oxatomide ≥99% (durch HPLC)
Catalog # ENZOBMLEI1140010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • VE:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :426.6 g/mol
Storage Temperature:Raumtemperatur
CAS Number:60607-34-3
Formula:C₂₇H₃₀N₄O
MDL Number:MFCD00211147