Order Entry
Austria
ContactUsLinkComponent
 
Trequinsin hydrochloride ≥98% (durch HPLC)
  ENZOBMLPD1800010
New Product
 :  
undefined
Trequinsin hydrochloride ≥98% (durch HPLC)
  ENZOBMLPD1800010
New Product
 :  

 

  • VE:
    10 mg
  • Seq:
    2275
  • Environmentally Preferable:

 

 

 :
 :405.5 · 36.5 g/mol
 :Raumtemperatur
 :78416-81-6
 :C₂₄H₂₇N₃O₃·HCl
 :MFCD01076563