Order Entry
Austria
ContactUsLinkComponent
Pharmaceutical Reference Standard, Acetaminophen Impurity
Pharmaceutical Reference Standard, Acetaminophen Impurity
Catalog # PROOTRC-A161235-25
CAS Number:  
Pharmaceutical Reference Standard, Acetaminophen Impurity
Catalog # PROOTRC-A161235-25
CAS Number:  

Specifications

  • VE:
    25 mg
  • CAS Nr.:
    98966-14-4
  • Bezeichnung:
    Acetaminophen Impurity
  • Shipping conditions:
    RT
  • Formel:
    C₁₆H₁₆N₂O₄
  • Lagertemperatur:
    4 °C
  • Molekulargewicht (Mw):
    300.31

Specifications

About this item