Rechargeable Lithium-ion Battery for M510/M610/M710 Printer
Catalog # BRDY175673
Supplier: Brady
Specifications
- Bezeichnung:Rechargeable lithium-ion battery for M510/M610/M710 printer, grey, 59,94W×94,50H×29,05D mm
- VE:1
Specifications
About this item
Work longer with this Li-ION M510, M610 and M710 battery pack. It enables cordless printer operation so you can print labels where (and when) you need them.
- Removable, rechargeable Li-ION Battery Pack
- Battery can be charged either while installed in an M610 or M710 printer, or while removed
- Prints up to 4,500 labels per charge
Work longer with this Li-ION M510, M610 and M710 Battery Pack. It enables cordless printer operation so you can print labels where (and when) you need them. Removable, rechargeable Li-ION Battery Pack. Battery can be charged either while installed in an M610 or M710 printer, or while removed. Prints up to 4,500 labels per charge.
Ordering information: Order reference M-LION-BATTERY.
Delivery: Li-Ion battery pack.