Crack Set for Nitrate Digestion
Catalog # WTWA250497
Supplier: WTW
CAS Number:
Specifications
- VE:90 Tests
- Bezeichnung:Crack set 20 for nitrate digestion for determination of total nitrogen
- Typ:Crack-SET 20 14963
- Inhalt:Reagent for 90 digestions
Specifications
About this item
Crack set 20 for Nitrate (14563, 14542 und 14773) for 90 determinations of total nitrogen in sample materials containing at most 15 mg/L N.