Order Entry
Austria
ContactUsLinkComponent
Sulbactam sodium, Pulver
Health
Sulbactam sodium, Pulver
Catalog # 1623671.
Supplier:  USP
CAS Number:  
Health
Sulbactam sodium, Pulver
Catalog # 1623671.
Supplier:  USP
CAS Number:  

Specifications

  • Packung:
    Vial
  • VE:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :255.223 g/mol
 :GHS08
CAS Number:69388-84-7
Formula:C₈H₁₀NO₅S·Na
MDL Number:MFCD01750374