Anti-Mouse Monoclonal Antibody [clone: [FluA-NP 4F1]] (HRP (Horseradish Peroxidase))
Catalog # PRSI99-735
Supplier: ProSci Inc.
Specifications
- Antikörper-Typ:Primär
- Klonalität:Monoklonal
- Klon:[FluA-NP 4F1]
- Konjugation:HRP (Horseradish Peroxidase)
- Durchflusszytometrie:Yes
- Wirt:Mouse
- Isotyp:IgG1
- Reaktivität:Virus
- Form:Yes
- Antigen-Synonyme:NP
- Lagerungspuffer:Supplied in a stock solution of 50% glycerol/50% PBS, pH 7,4; No preservative added
- Lagertemperatur:Store vial at 2...8 °C
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant influenza virus type A nucleoprotein
- Packung:Vial
- VE:1 mL
Specifications
About this item
Type: Primary
Antigen:
Clonality: Monoclonal
Clone: [FluA-NP 4F1]
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus