Order Entry
Austria
ContactUsLinkComponent
 
Anti-GluR3 Rabbit Monoclonal Antibody [clone: D25G9]
  CSTA5117S
Anti-GluR3 Rabbit Monoclonal Antibody [clone: D25G9]
  CSTA5117S

 

  • Antikörper-Typ:
    Primär
  • Antigen-Symbol:
    GluR3
  • Klonalität:
    Monoklonal
  • Klon:
    D25G9
  • Wirt:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Sequence:
    CLYNTNQNTTEKPFHLNYHVDHLDSS
  • Lagertemperatur:
    –20 °C
  • Transporttemperatur:
    Ambient
  • VE:
    100 µl

 

 

Type: Primary
Antigen: GluR3
Clonality: Monoclonal
Clone: D25G9
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat