Anti-DIAP1 Rabbit Polyclonal Antibody
CSTA5486S
- Antikörper-Typ:Primär
- Antigen-Symbol:DIAP1
- Klonalität:Polyklonal
- Wirt:Rabbit
- Reaktivität:Monkey,Human
- Western blot:Yes
- Sequence:CNSETFPTILEEAKELVGRAS
- Lagertemperatur:−20 °C
- Transporttemperatur:Ambient
- VE:100 µl
Type: Primary
Antigen: DIAP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Monkey