Anti-PASK Rabbit Monoclonal Antibody [clone: C70B2]
Catalog # CSTA3086S
Supplier: Cell Signaling
Specifications
- Antikörper-Typ:Primär
- Antigen-Symbol:PASK
- Klonalität:Monoklonal
- Klon:C70B2
- Wirt:Rabbit
- Immunpräzipitation:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CGQHIPKSLKIQRSVGRARDGTT
- Lagertemperatur:−20 °C
- Transporttemperatur:Ambient
- VE:100 µl
Specifications
About this item
Type: Primary
Antigen: PASK
Clonality: Monoclonal
Clone: C70B2
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat