Rat IgA ELISA Kit
Artikelnr. BMOLE111-102
Lieferant: BIOMOL RESEARCH LABORATORIES
CAS-Nummer:
Spezifikationen
- VE:1 KIT
- Format:Sachets
- Bezeichnung:Rat IgA ELISA kit
- Lagertemperatur:4 °C
- Anwendung:ELISA
- Transporttemperatur:4 °C (International: 4 °C)
Spezifikationen
Über diesen Artikel
This ELISA is for the detection of rat IgA in serum or plasma. Due to idiotypic and allotypic variations, this product may not accurately quantitate hybridomas, monoclonals or myelomas. This kit contains sufficient components to quantitate rat IgA in up to 40 samples, tested in duplicate.