D(-)-Ribose
Glass bottle for solids, 100 g
MOLE13992794-100G
Marketsource
: Molekula
- :50-69-1
- :C₅H₁₀O₅
- :MFCD00135453
- :150.13 g/mol
- :12,08371
- :331 °C (1013 hPa)
- :90…95 °C
- :0,8 g/cm³ (20 °C)
- :Raumtemperatur
- :200-059-4
- Packung:Glass bottle for solids
- VE:100 g