HIV virus, model
AIDS virus model
Artikelnr. 763-0132
Marketsource
Anbieter: 3B Scientific
Springe zu:
Product Details & Documents
Model of HIV virus enlarged 1 million. times. The model shows the different parts of the virus, including the RNA complex. Height 18 cm. Diam. 12 cm.
Spezifikationen
- Bezeichnung:AIDS virus model