Native Megathura crenulata KLH (from Megathura crenulata), Chloramphenicol
AATB50002
:
- Packung:Vial
- VE:1 mg
- Konjugation:Chloramphenicol
- Protein/Peptid-Typ:Nativ
- Ursprung:Megathura crenulata
- Spezies:Megathura crenulata
- Proteinsynonym:Keyhole Limpet Hemocyanin
- Protein/Peptid-Name:KLH
Chloramphenicol-KLH conjugate is used for developing anti-chloramphenicol antibodies that are typically used in an ELISA assay.
- Antibody development
- High immunogenicity
: For research use only.