ContactUsLinkComponent
Native Megathura crenulata KLH (from Megathura crenulata), Chloramphenicol
  AATB50002
 :  
undefined
Native Megathura crenulata KLH (from Megathura crenulata), Chloramphenicol
  AATB50002
 :  
  • Packung:
    Vial
  • VE:
    1 mg
  • Konjugation:
    Chloramphenicol
  • Protein/Peptid-Typ:
    Nativ
  • Ursprung:
    Megathura crenulata
  • Spezies:
    Megathura crenulata
  • Proteinsynonym:
    Keyhole Limpet Hemocyanin
  • Protein/Peptid-Name:
    KLH
Chloramphenicol-KLH conjugate is used for developing anti-chloramphenicol antibodies that are typically used in an ELISA assay.
  • Antibody development
  • High immunogenicity
 : For research use only.