ContactUsLinkComponent
ReadiLink™ Rapid mFluor™ Violet 420 Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
ReadiLink™ Rapid mFluor™ Violet 420 Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Catalog # AATB1105
Supplier:  AAT Bioquest
CAS Number:  
undefined
ReadiLink™ Rapid mFluor™ Violet 420 Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Catalog # AATB1105
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • VE:
    2 Tests
  • Bezeichnung:
    ReadiLink™ rapid mFluor™ violet 420 antibody labelling kit 'Microscale optimized for labelling 50 µg antibody per reaction'
Specifications
About this item
AAT Bioquest's mFluor™ dyes are developed for flow cytometry-focused applications.
  • Violet-laser excitable
  • Robust and simple antibody labelling protocol
  • No column purification required
Caution: For research use only.