ReadiLink™Rapid mFluor™ Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Catalog # AATB1131
Supplier: AAT Bioquest
CAS Number:
Specifications
- VE:2 Tests
- Bezeichnung:ReadiLink™ rapid mFluor™ red 780 antibody labelling kit 'Microscale optimized for labelling 50 µg antibody per reaction'
Specifications
About this item
mFluor™ dyes are developed for flow cytometry-focused applications.
- Red-laser excitable
- Robust and simple antibody labelling protocol
- No column purification required
Caution: For research use only.