ReadiLink™ Rapid mFluor™ Green 620 Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Artikelnr. AATB1123
Lieferant: AAT Bioquest
CAS-Nummer:
Spezifikationen
- VE:2 Tests
- Bezeichnung:ReadiLink™ rapid mFluor™ green 620 antibody labelling kit 'Microscale optimized for labelling 50 µg antibody per reaction'
Spezifikationen
Über diesen Artikel
AAT Bioquest's mFluor™ dyes are developed for flow cytometry-focused applications.
- Green-laser excitable
- Robust and simple antibody labelling protocol
- No column purification required
Achtung! Bitte beachten: For research use only.