Amplite® Colorimetric α-Ketoglutarate Quantitation Kit
Catalog # AATB10085
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- VE:200 Tests
- Bezeichnung:Amplite™ colorimetric α-ketoglutarate quantitation kit
Specifications
About this item
Alpha-ketoglutarate (α-Ketoglutarate) is a key molecule in the Krebs cycle determining the overall rate of the citric acid cycle of the organism.
- In-house QC testing
- Sensitive
- Suggested protocol available
This colorimetric α-Ketoglutarate Quantitation Kit offers a sensitive colorimetric assay for quantifying α-ketoglutarate in biological samples.
Caution: For research use only.