Buccutite™ Rapid Protein Crosslinking Kit *Microscale Optimized for Crosslinking 100 ug Antibody Per Reaction* (2 Labelings)
Catalog # AATB1315
Supplier: AAT Bioquest
CAS Number:
Specifications
- VE:2 Tests
- Bezeichnung:Buccutite™ rapid protein crosslinking kit 'Microscale optimized for crosslinking 100 µg antibody per reaction'
Specifications
About this item
Protein-protein conjugations are commonly performed with a bifunctional linker such as SMCC.
- No catalyst required
- Neutral crosslinking
- Robust and simple protocol
Caution: For research use only.