ContactUsLinkComponent
PSA™ iFluor® 647 Imaging Kit with Goat Anti-Mouse IgG
PSA™ iFluor® 647 Imaging Kit with Goat Anti-Mouse IgG
Catalog # AATB45290
Supplier:  AAT Bioquest
CAS Number:  
undefined
PSA™ iFluor® 647 Imaging Kit with Goat Anti-Mouse IgG
Catalog # AATB45290
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • VE:
    100 Tests
  • Bezeichnung:
    Can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10 - 50 times higher than the widely used tyramide (TSA) reagents
Specifications
About this item
iFluor® 647 PSA kit is a much superior replacement for Alexa Fluor 647 tyramide-based kit or other spectrally similar fluorescent tyramide or TSA kits.
  • Higher efficiency
  • Improved fluorescence signal
  • Less consumption of primary antibody
  • 10 to 50X improvement over TSA reagents
In combination with our superior iFluor® dyes that have higher florescence intensity, increased photostability and enhanced water solubility, the iFluor® dye-labeled Styramide™ conjugates can generate fluorescence signal with significantly higher precision and sensitivity (more than 100 times) than standard ICC/IF/IHC.
Packaging: Plastic vial.
Caution: For research use only.