ReadiLink™ Rapid iFluor® 594 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Artikelnr. AATB1230
Lieferant: AAT Bioquest
CAS-Nummer:
Spezifikationen
- VE:2 Tests
- Bezeichnung:ReadiLink™ rapid iFluor™ 594 antibody labeling kit 'Microscale optimized for labeling 50 µg antibody per reaction'
Spezifikationen
Über diesen Artikel
AAT Bioquest's iFluor® dyes are optimised for labelling proteins, in particular, antibodies.
- Red-laser excitable
- Robust and simple antibody labelling protocol
- No column purification required
Achtung! Bitte beachten: For research use only.