PhosphoWorks™ Colorimetric Phosphate Assay Kit, Blue Color
AATB21665
:
- VE:1.000 Tests
- Bezeichnung:PhosphoWorks™ colorimetric phosphate assay kit *blue color*
- Tests pro Kit:1000 Tests
Phosphate is involved in many biological reactions.
- Stable end-point signal
- Multiple applications
- Photometric detection
: For research use only.