MitoLite™ Red FX600
MitoLite™ Red, 500 Tests, 500 Tests
Artikelnr. AATB22677
Anbieter: AAT Bioquest
Springe zu:
Product Details & Documents
MitoLite™ Red is a cationic dye that selectively accumulates in mitochondria probably vial the mitochondrial membrane potential gradient.
- Mitochondria-Specific Dye
- Cell-Permeable
- Robust
: For research use only.
Spezifikationen
Spezifikationen
- VE:500 Tests
- Bezeichnung:MitoLite™ Red
- Tests pro Kit:500 Tests
- Cat. No.:AATB22677
- Purity:95%