Tide Fluor™ 2WS succinimidyl ester, Tide Fluor™ fluoreszierende Probe
Catalog # AATB2349
Supplier: AAT Bioquest
CAS Number:
Specifications
- Packung:Plastic bottle
- VE:5 mg
- Seq:2210
Specifications
About this item
:
:945.07 g/mol
ADR:5,II
:GHS07