VWR® Low Actinic Volumetric Flasks with PE Stoppers, Amber Glass, Class A, Serialised
Lieferant: VWR Collection
Zertifikate
Über diesen Artikel
Class A, serialiased, wide-mouth, low actinic volumetric flasks are supplied with polyethylene stoppers.
- Wide mouth
- Amber glass fabrication
- Polyethylene stoppers
Ordering information: Flasks are individually certified.
Frequently Bought Together