Order Entry
Austria
ContactUsLinkComponent
Recombinant Human TNFRSF10B (from Baculovirus), hIgG-His-tag
Recombinant Human TNFRSF10B (from Baculovirus), hIgG-His-tag
Catalog # ABNOP8039.50UG
Supplier:  Abnova
CAS Number:  
Recombinant Human TNFRSF10B (from Baculovirus), hIgG-His-tag
Catalog # ABNOP8039.50UG
Supplier:  Abnova
CAS Number:  

Specifications

  • VE:
    50 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    Baculovirus
  • Spezies:
    Human
  • Tag sequence:
    hIgG-His-tag
  • Gen-ID:
    O14763
  • Protein/Peptid-Name:
    TNFRSF10B
  • Sequence:
    56a.a.-210a.a.

Specifications

About this item