Order Entry
Austria
ContactUsLinkComponent
Recombinant Mouse CST3 (from Baculovirus), His-tag
Recombinant Mouse CST3 (from Baculovirus), His-tag
Catalog # ABNOP8018.100UG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse CST3 (from Baculovirus), His-tag
Catalog # ABNOP8018.100UG
Supplier:  Abnova
CAS Number:  

Specifications

  • VE:
    100 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    Baculovirus
  • Spezies:
    Mouse
  • Tag sequence:
    His-tag
  • Gen-ID:
    P21460
  • Protein/Peptid-Name:
    CST3
  • Sequence:
    21a.a.-140a.a.

Specifications

About this item