Recombinant Mouse TNF (from Baculovirus), His-tag
ABNOP7950.100UG
: Abnova
:
- VE:100 µG
- Protein/Peptid-Typ:Rekombinant
- Ursprung:Baculovirus
- Spezies:Mouse
- Tag sequence:His-tag
- Gen-ID:P06804
- Protein/Peptid-Name:TNF
- Sequence:8a.a.-235a.a.