Order Entry
Austria
ContactUsLinkComponent
 
Recombinant Mouse P4HB (from Baculovirus), His-tag
  ABNOP7971.250UG
 :  Abnova
 :  
Recombinant Mouse P4HB (from Baculovirus), His-tag
  ABNOP7971.250UG
 :  Abnova
 :  

 

  • VE:
    250 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    Baculovirus
  • Spezies:
    Mouse
  • Tag sequence:
    His-tag
  • Gen-ID:
    P09103
  • Protein/Peptid-Name:
    P4HB
  • Sequence:
    20a.a.-509a.a.